Assay Details
Gene Symbol: As3mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080890.1 | NP_543166.1 | 9 - 10 | 919 | 102 | |
GenBank mRNA | AF393243.1 | - | 9 - 10 | 919 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein methylation
toxin metabolic process
arsonoacetate metabolic process
methylation
response to arsenic-containing substance
protein methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
arsenite methyltransferase activity
methylarsonite methyltransferase activity
protein methylation
toxin metabolic process
arsonoacetate metabolic process
methylation
response to arsenic-containing substance
protein methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
arsenite methyltransferase activity
methylarsonite methyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P13091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7286 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13091 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13091 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P13091 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P372419 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13091 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95679 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63916 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51529 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10715880 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715887 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715888 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715876 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715879 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715883 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715881 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715884 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715878 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715877 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715886 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715885 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10715875 | - | - |
Rat Genome 230 2.0 Array | 1368244_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368244_at | - | - |
Affymetrix HT Rat Focus Array | 1368244_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368244_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100030670.rn.1 | TC0100003752.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100030669.rn.1 | TC0100003752.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100030667.rn.1 | TC0100003752.rn.1 | RTA TC |
Rat Expression Set 230 | 1368244_at | - | - |
Rat Expression Set 230 | 1368244_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10715875 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21640156 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349101 | - | - |
RatRef-12 Expression BeadChip | ILMN_50501 | - | - |
