Assay Details
Gene Symbol: Hadhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239786.1 | XP_006239848.1 | 4 - 5 | 688 | 76 | |
XM_006239788.1 | XP_006239850.1 | 4 - 5 | 344 | 76 | ||
XM_006239789.2 | XP_006239851.1 | 4 - 5 | 619 | 76 | ||
GenBank mRNA | BC060545.1 | - | 4 - 5 | 271 | 6918791 | 76 |
D16479.1 | - | 3 - 4 | 235 | 76 | ||
FM050540.1 | - | 4 - 5 | 299 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
