Assay Details
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134459.1 | NP_604454.1 | 7 - 8 | 525 | 67 | |
XM_006251676.2 | XP_006251738.1 | 7 - 8 | 561 | 67 | ||
XM_008770596.1 | XP_008768818.1 | 7 - 8 | 551 | 67 | ||
GenBank mRNA | AB031014.1 | - | 7 - 8 | 525 | 67 | |
AF481858.1 | - | 7 - 8 | 472 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
