Assay Details
Gene Symbol: Ccnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC059113.1 | - | 1 - 2 | 118 | 85 | |
FN800625.1 | - | 1 - 2 | 116 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC059113.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
