Assay Details
Gene Symbol: Elavl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238385.2 | XP_006238447.1 | 4 - 5 | 557 | 77 | |
XM_006238388.2 | XP_006238450.1 | 3 - 4 | 360 | 77 | ||
XM_006238389.2 | XP_006238451.1 | 3 - 4 | 897 | 77 | ||
XM_006238391.2 | 3 - 4 | 897 | 77 | |||
XM_006238395.2 | XP_006238457.1 | 2 - 3 | 288 | 77 | ||
XM_008763848.1 | XP_008762070.1 | 2 - 3 | 603 | 77 | ||
XM_008763849.1 | XP_008762071.1 | 2 - 3 | 603 | 77 | ||
XM_008763850.1 | XP_008762072.1 | 2 - 3 | 266 | 77 | ||
GenBank mRNA | AB098713.1 | - | 3 - 4 | 821 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
