Assay Details
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019377.1 | NP_062250.1 | 2 - 3 | 463 | 79 | |
GenBank mRNA | BC076502.1 | - | 2 - 3 | 457 | 79 | |
D17446.1 | - | 2 - 3 | 458 | 79 | ||
FQ214109.1 | - | 2 - 3 | 499 | 79 | ||
FQ229978.1 | - | 2 - 3 | 468 | 79 | ||
S55223.1 | - | 2 - 3 | 458 | 79 | ||
S83440.1 | - | 2 - 3 | 463 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
