Assay Details
Gene Symbol: Ndufaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020080.2 | NP_064465.1 | 1 - 2 | 764 | 70 | |
XM_006243845.2 | XP_006243907.1 | 2 - 3 | 2246 | 70 | ||
GenBank mRNA | FQ216095.1 | - | 1 - 2 | 115 | 70 | |
U95160.1 | - | 1 - 2 | 100 | 70 | ||
U95161.1 | - | 1 - 2 | 100 | 70 | ||
U95162.1 | - | 1 - 2 | 100 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
