Assay Details
Gene Symbol: Dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022389.2 | NP_071784.1 | 2 - 3 | 320 | 117 | |
XM_008760171.1 | XP_008758393.1 | 1 - 2 | 715 | 117 | ||
GenBank mRNA | AF272393.1 | - | 2 - 3 | 320 | 117 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF272393.1, NM_022389.2, XM_008760171.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
