Assay Details
Gene Symbol: Pja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277278.1 | NP_001264207.1 | 5 - 6 | 1664 | 113 | |
NM_138896.2 | NP_620251.1 | 6 - 7 | 1850 | 113 | ||
GenBank mRNA | BC074015.1 | - | 6 - 7 | 1775 | 113 | |
D32249.1 | - | 6 - 7 | 1786 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
