Assay Details
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012801.1 | NP_036933.1 | 3 - 4 | 552 | 74 | |
XM_006248918.2 | XP_006248980.1 | 1 - 2 | 534 | 74 | ||
XM_006248919.2 | XP_006248981.1 | 1 - 2 | 534 | 74 | ||
XM_008768940.1 | XP_008767162.1 | 2 - 3 | 2133 | 74 | ||
GenBank mRNA | BC061731.1 | - | 1 - 2 | 539 | 74 | |
D10106.1 | - | 1 - 2 | 111 | 74 | ||
L06238.1 | - | 3 - 4 | 552 | 74 | ||
L06894.1 | - | 2 - 3 | 196 | 74 | ||
Z14120.1 | - | 1 - 2 | 46 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
