Assay Details
Gene Symbol: Fgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020071.2 | NP_064456.2 | 1 - 2 | 120 | 87 | |
GenBank mRNA | BC088102.1 | - | 1 - 2 | 120 | 87 | |
U05675.1 | - | 1 - 2 | 121 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
