Assay Details
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031037.2 | NP_112299.1 | 5 - 6 | 657 | 81 | |
GenBank mRNA | AF277892.1 | - | 4 - 5 | 438 | 81 | |
AF397193.1 | - | 1 - 2 | 237 | 81 | ||
BC065579.1 | - | 5 - 6 | 657 | 81 | ||
DQ120485.1 | - | 4 - 5 | 422 | 81 | ||
DQ120486.1 | - | 4 - 5 | 422 | 81 | ||
FQ209560.1 | - | 1 - 2 | 45 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
