Assay Details
Gene Symbol: Sqstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175843.4 | NP_787037.2 | 4 - 5 | 697 | 103 | |
XM_006246214.2 | XP_006246276.1 | 4 - 5 | 719 | 103 | ||
GenBank mRNA | BC061575.1 | - | 4 - 5 | 701 | 6887571 | 103 |
Y08355.1 | - | 4 - 5 | 726 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
