Assay Details
Gene Symbol: Sqstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175843.4 | NP_787037.2 | 5 - 6 | 773 | 113 | |
XM_006246214.2 | XP_006246276.1 | 5 - 6 | 795 | 113 | ||
GenBank mRNA | BC061575.1 | - | 5 - 6 | 777 | 6887571 | 113 |
Y08355.1 | - | 5 - 6 | 802 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
