Assay Details
Gene Symbol: Sqstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175843.4 | NP_787037.2 | 6 - 7 | 998 | 71 | |
XM_006246213.2 | XP_006246275.1 | 5 - 6 | 939 | 71 | ||
XM_006246214.2 | XP_006246276.1 | 6 - 7 | 1020 | 71 | ||
XM_006246215.2 | XP_006246277.1 | 5 - 6 | 939 | 71 | ||
GenBank mRNA | BC061575.1 | - | 6 - 7 | 1002 | 6887571 | 71 |
FQ235251.1 | - | 1 - 2 | 141 | 71 | ||
Y08355.1 | - | 6 - 7 | 1027 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
