Assay Details
Gene Symbol: Mapkapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178102.2 | NP_835203.1 | 3 - 4 | 679 | 67 | |
GenBank mRNA | AY197741.1 | - | 3 - 4 | 679 | 67 | |
BC062048.1 | - | 3 - 4 | 645 | 5598069 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
