Assay Details
Gene Symbol: Cyp27a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245175.1 | XP_006245237.1 | 6 - 7 | 1070 | 64 | |
GenBank mRNA | BC061848.1 | - | 6 - 7 | 1234 | 5621918 | 64 |
M38566.1 | - | 6 - 7 | 1235 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
