Assay Details
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082477.2 | NP_001075946.2 | 1 - 2 | 43 | 77 | |
NM_178866.4 | NP_849197.3 | 1 - 2 | 43 | 77 | ||
GenBank mRNA | BC086374.1 | - | 1 - 2 | 43 | 77 | |
M15480.1 | - | 1 - 2 | 87 | 77 | ||
M15481.1 | - | 1 - 2 | 775 | 77 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC086374.1, M15480.1, M15481.1, NM_001082477.2, NM_178866.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
