Assay Details
Gene Symbol: Tnfsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181384.2 | NP_852049.1 | 1 - 2 | 522 | 70 | |
GenBank mRNA | AY259541.1 | - | 1 - 2 | 522 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
