Assay Details
Gene Symbol: Tfb1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181474.2 | NP_852139.2 | 5 - 6 | 688 | 75 | |
GenBank mRNA | AY198392.1 | - | 5 - 6 | 668 | 75 | |
BC070907.1 | - | 5 - 6 | 688 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
