Assay Details
Gene Symbol: LOC103689957 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239732.2 | XP_006239794.1 | 1 - 2 | 1916 | 64 | |
XM_006239733.2 | XP_006239795.1 | 1 - 2 | 1916 | 64 |
Gene Symbol: Fbxo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181631.2 | NP_853662.1 | 1 - 2 | 132 | 64 | |
XM_008764477.1 | XP_008762699.1 | 1 - 2 | 1909 | 64 | ||
GenBank mRNA | AY274810.1 | - | 1 - 2 | 132 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
