Assay Details
Gene Symbol: Samd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107498.1 | NP_001100968.1 | 1 - 2 | 403 | 90 | |
XM_008759143.1 | XP_008757365.1 | 2 - 3 | 526 | 90 | ||
GenBank mRNA | BC090319.1 | - | 2 - 3 | 493 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
