Assay Details
Gene Symbol: Samd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107498.1 | NP_001100968.1 | 2 - 3 | 872 | 64 | |
XM_008759143.1 | XP_008757365.1 | 3 - 4 | 995 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
