Assay Details
Gene Symbol: Tp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030989.3 | NP_112251.2 | 5 - 6 | 682 | 67 | |
XM_006246594.2 | XP_006246656.1 | 6 - 7 | 1111 | 67 | ||
XM_006246595.2 | XP_006246657.1 | 5 - 6 | 1468 | 67 | ||
XM_008767773.1 | XP_008765995.1 | 5 - 6 | 689 | 67 | ||
GenBank mRNA | AF209191.1 | - | 4 - 5 | 574 | 67 | |
BC081788.1 | - | 5 - 6 | 611 | 67 | ||
BC098663.1 | - | 5 - 6 | 640 | 67 | ||
FQ232659.1 | - | 5 - 6 | 666 | 67 | ||
U90328.1 | - | 4 - 5 | 551 | 67 | ||
X13058.1 | - | 4 - 5 | 574 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
