Assay Details
Gene Symbol: Camk1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182842.1 | NP_878262.1 | 4 - 5 | 377 | 73 | |
XM_006250470.2 | XP_006250532.1 | 4 - 5 | 494 | 73 | ||
GenBank mRNA | AB101231.1 | - | 4 - 5 | 377 | 73 | |
AB101232.1 | - | 4 - 5 | 377 | 73 | ||
D86557.1 | - | 4 - 5 | 383 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
