Assay Details
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183327.1 | NP_899156.1 | 1 - 2 | 109 | 102 | |
XM_006246125.2 | XP_006246187.1 | 1 - 2 | 672 | 102 | ||
GenBank mRNA | AY574252.1 | - | 1 - 2 | 109 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
