Assay Details
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183327.1 | NP_899156.1 | 8 - 9 | 1129 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chloride transport
signal transduction
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
post-embryonic development
adult behavior
synaptic transmission, GABAergic
cellular response to histamine
chloride transmembrane transport
GABA-A receptor activity
inhibitory extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
GABA receptor activity
chloride transport
signal transduction
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
post-embryonic development
adult behavior
synaptic transmission, GABAergic
cellular response to histamine
chloride transmembrane transport
GABA-A receptor activity
inhibitory extracellular ligand-gated ion channel activity
chloride channel activity
protein binding
GABA receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P486909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143494 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P486909 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P486909 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P486909 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P486909 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88902 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37744 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66741 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84474 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61488 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51667 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68049 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50876 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10742168 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742165 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742164 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742169 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742167 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742166 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742162 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742163 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742170 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10742160 | - | - |
Rat Genome 230 2.0 Array | 1391653_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391653_at | - | - |
Affymetrix HT Rat Focus Array | 1391653_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1391653_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000023227.rn.1 | TC1000002598.rn.1 | RTA TC |
Rat Expression Set 230 | 1391653_at | - | - |
Rat Expression Set 230 | 1391653_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10742160 | - | - |
Rat Genome U34 Set | L08497cds_at | - | - |
Rat Genome U34 Set | L08497cds_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20707933 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369778 | - | - |
RatRef-12 Expression BeadChip | ILMN_56631 | - | - |
