Assay Details
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_184046.1 | NP_908935.1 | 654 | 89 | ||
XM_008763055.1 | XP_008761277.1 | 446 | 89 | |||
GenBank mRNA | AY348867.1 | - | 654 | 89 | ||
AY348868.1 | - | 654 | 89 |
Gene Symbol: LOC103692165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763036.1 | XP_008761258.1 | 1 - 2 | 446 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
