Assay Details
Gene Symbol: Rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_184046.1 | NP_908935.1 | 1090 | 66 | ||
XM_006236729.2 | XP_006236791.1 | 3118 | 66 | |||
XM_006236732.2 | XP_006236794.1 | 3118 | 66 | |||
XM_008763054.1 | XP_008761276.1 | 1045 | 66 | |||
XM_008763055.1 | XP_008761277.1 | 882 | 66 | |||
GenBank mRNA | AY348867.1 | - | 1090 | 66 |
Gene Symbol: LOC103692165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763034.1 | XP_008761256.1 | 5 - 6 | 3118 | 66 | |
XM_008763035.1 | XP_008761257.1 | 5 - 6 | 1045 | 66 | ||
XM_008763036.1 | XP_008761258.1 | 5 - 6 | 882 | 66 | ||
XM_008763038.1 | XP_008761260.1 | 5 - 6 | 3118 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
