Assay Details
Gene Symbol: Ppp2r5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106740.1 | NP_001100210.1 | 2 - 3 | 688 | 133 | |
XM_006240222.2 | XP_006240284.1 | 2 - 3 | 738 | 133 | ||
XM_006240223.2 | XP_006240285.1 | 2 - 3 | 736 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
