Assay Details
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031643.4 | NP_113831.1 | 10 - 11 | 1113 | 130 | |
GenBank mRNA | BC089772.1 | - | 10 - 11 | 1107 | 7382030 | 130 |
D13341.1 | - | 10 - 11 | 1096 | 130 | ||
D14591.1 | - | 10 - 11 | 1066 | 130 | ||
X62313.1 | - | 10 - 11 | 1103 | 130 | ||
Z16415.1 | - | 10 - 11 | 1096 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
regulation of vascular smooth muscle contraction
protein phosphorylation
response to oxidative stress
cell cycle arrest
mitotic nuclear division
cell proliferation
negative regulation of cell proliferation
positive regulation of gene expression
peptidyl-tyrosine phosphorylation
neuron differentiation
keratinocyte differentiation
positive regulation of cell migration
melanosome transport
regulation of stress-activated MAPK cascade
positive regulation of transcription elongation from RNA polymerase II promoter
negative regulation of homotypic cell-cell adhesion
peptidyl-tyrosine dephosphorylation
intracellular signal transduction
positive regulation of GTPase activity
positive regulation of cell differentiation
positive regulation of Ras protein signal transduction
vesicle transport along microtubule
Golgi inheritance
response to axon injury
regulation of axon regeneration
neuron projection morphogenesis
cell motility
positive regulation of axonogenesis
protein heterooligomerization
response to glucocorticoid
lung morphogenesis
trachea formation
epithelial cell proliferation involved in lung morphogenesis
placenta blood vessel development
labyrinthine layer development
ERK1 and ERK2 cascade
positive regulation of protein serine/threonine kinase activity
regulation of Golgi inheritance
cellular senescence
regulation of early endosome to late endosome transport
protein serine/threonine kinase activity
MAP kinase kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
receptor signaling protein tyrosine phosphatase activity
protein binding
ATP binding
Ras GTPase binding
protein kinase binding
mitogen-activated protein kinase kinase kinase binding
protein serine/threonine kinase activator activity
MAPK cascade
activation of MAPK activity
regulation of vascular smooth muscle contraction
protein phosphorylation
response to oxidative stress
cell cycle arrest
mitotic nuclear division
cell proliferation
negative regulation of cell proliferation
positive regulation of gene expression
peptidyl-tyrosine phosphorylation
neuron differentiation
keratinocyte differentiation
positive regulation of cell migration
melanosome transport
regulation of stress-activated MAPK cascade
positive regulation of transcription elongation from RNA polymerase II promoter
negative regulation of homotypic cell-cell adhesion
peptidyl-tyrosine dephosphorylation
intracellular signal transduction
positive regulation of GTPase activity
positive regulation of cell differentiation
positive regulation of Ras protein signal transduction
vesicle transport along microtubule
Golgi inheritance
response to axon injury
regulation of axon regeneration
neuron projection morphogenesis
cell motility
positive regulation of axonogenesis
protein heterooligomerization
response to glucocorticoid
lung morphogenesis
trachea formation
epithelial cell proliferation involved in lung morphogenesis
placenta blood vessel development
labyrinthine layer development
ERK1 and ERK2 cascade
positive regulation of protein serine/threonine kinase activity
regulation of Golgi inheritance
cellular senescence
regulation of early endosome to late endosome transport
protein serine/threonine kinase activity
MAP kinase kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
receptor signaling protein tyrosine phosphatase activity
protein binding
ATP binding
Ras GTPase binding
protein kinase binding
mitogen-activated protein kinase kinase kinase binding
protein serine/threonine kinase activator activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P531063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113135 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P531063 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P531063 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P531063 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P531063 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48070 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18162 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5814 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54976 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103201 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10918220 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918213 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918215 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918219 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918212 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918210 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918211 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918221 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918216 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918214 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918217 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918218 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10918209 | - | - |
Rat Genome 230 2.0 Array | 1367760_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367760_at | - | - |
Affymetrix HT Rat Focus Array | 1367760_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367760_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800022333.rn.1 | TC0800002747.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800022332.rn.1 | TC0800002747.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800022334.rn.1 | TC0800002747.rn.1 | RTA TC |
Rat Expression Set 230 | 1367760_PM_at | - | - |
Rat Expression Set 230 | 1367760_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918209 | - | - |
Rat Genome U34 Set | D14591_s_at | - | - |
Rat Genome U34 Set | L04485mRNA_s_at | - | - |
Rat Genome U34 Set | rc_AI178835_at | - | - |
Rat Genome U34 Set | D14591_s_at | - | - |
Rat Genome U34 Set | L04485mRNA_s_at | - | - |
Rat Genome U34 Set | rc_AI178835_at | - | - |
Rat Genome U34 Set | rc_AI178835_at | - | - |
Rat Genome U34 Set | D14591_s_at | - | - |
Rat Genome U34 Set | L04485mRNA_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21080101 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361975 | - | - |
RatRef-12 Expression BeadChip | ILMN_59171 | - | - |
