Assay Details
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012554.3 | NP_036686.2 | 1 - 2 | 114 | 80 | |
XM_006239444.1 | XP_006239506.1 | 1 - 2 | 130 | 80 | ||
GenBank mRNA | BC063174.1 | - | 1 - 2 | 107 | 80 | |
BC081847.1 | - | 1 - 2 | 87 | 7189453 | 80 | |
BC091572.1 | - | 1 - 2 | 84 | 7107492 | 80 | |
BC097343.1 | - | 1 - 2 | 80 | 7461555 | 80 | |
CB809779.1 | - | 1 - 2 | 114 | 80 | ||
FQ225935.1 | - | 1 - 2 | 127 | 80 | ||
X02610.1 | - | 1 - 2 | 103 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC081847.1,BC091572.1,BC097343.1,NM_012554.3,X02610.1,XM_006239444.1 transcripts.
This assay is in the 5'UTR region of the BC081847.1,BC091572.1,BC097343.1,NM_012554.3,X02610.1,XM_006239444.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
