Assay Details
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109908.1 | NP_001103378.1 | 3 - 4 | 354 | 85 | |
NM_012554.3 | NP_036686.2 | 3 - 4 | 305 | 85 | ||
GenBank mRNA | BC063174.1 | - | 3 - 4 | 298 | 85 | |
BC078896.1 | - | 3 - 4 | 354 | 7112523 | 85 | |
BC081847.1 | - | 3 - 4 | 278 | 7189453 | 85 | |
BC090069.1 | - | 3 - 4 | 235 | 7108936 | 85 | |
BC091572.1 | - | 3 - 4 | 275 | 7107492 | 85 | |
BC097343.1 | - | 3 - 4 | 271 | 7461555 | 85 | |
CB809779.1 | - | 3 - 4 | 305 | 85 | ||
X02610.1 | - | 3 - 4 | 294 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
