Assay Details
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013006.1 | NP_037138.1 | 3 - 4 | 244 | 129 | |
XM_008763522.1 | XP_008761744.1 | 2 - 3 | 236 | 129 | ||
GenBank mRNA | BC085750.1 | - | 3 - 4 | 182 | 7190638 | 129 |
D63885.1 | - | 3 - 4 | 244 | 129 | ||
U97146.1 | - | 3 - 4 | 272 | 129 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_008763522.1 transcripts.
This assay is in the 5'UTR region of the XM_008763522.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
