Assay Details
Gene Symbol: Cfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017147.2 | NP_058843.1 | 2 - 3 | 384 | 90 | |
GenBank mRNA | BC059143.1 | - | 2 - 3 | 384 | 90 | |
BC086533.1 | - | 2 - 3 | 369 | 90 | ||
FQ212136.1 | - | 1 - 2 | 256 | 90 | ||
FQ212794.1 | - | 2 - 3 | 383 | 90 | ||
FQ212910.1 | - | 2 - 3 | 379 | 90 | ||
FQ213701.1 | - | 2 - 3 | 379 | 90 | ||
FQ213897.1 | - | 2 - 3 | 408 | 90 | ||
FQ214120.1 | - | 2 - 3 | 380 | 90 | ||
FQ214438.1 | - | 2 - 3 | 379 | 90 | ||
FQ220384.1 | - | 2 - 3 | 395 | 90 | ||
FQ220571.1 | - | 2 - 3 | 381 | 90 | ||
FQ226405.1 | - | 2 - 3 | 379 | 90 | ||
FQ227487.1 | - | 2 - 3 | 381 | 90 | ||
FQ229073.1 | - | 2 - 3 | 408 | 90 | ||
FQ229766.1 | - | 2 - 3 | 383 | 90 | ||
FQ229833.1 | - | 2 - 3 | 379 | 90 | ||
FQ229875.1 | - | 2 - 3 | 383 | 90 | ||
FQ230001.1 | - | 2 - 3 | 383 | 90 | ||
FQ230091.1 | - | 2 - 3 | 380 | 90 | ||
FQ231379.1 | - | 2 - 3 | 379 | 90 | ||
FQ232577.1 | - | 1 - 2 | 259 | 90 | ||
FQ233750.1 | - | 2 - 3 | 379 | 90 | ||
FQ234162.1 | - | 2 - 3 | 383 | 90 | ||
FQ234303.1 | - | 2 - 3 | 379 | 90 | ||
X62908.1 | - | 1 - 2 | 308 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
