Assay Details
Gene Symbol: Pebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017236.1 | NP_058932.1 | 3 - 4 | 406 | 67 | |
GenBank mRNA | BC063171.1 | - | 3 - 4 | 423 | 67 | |
FQ211637.1 | - | 3 - 4 | 367 | 67 | ||
FQ212424.1 | - | 3 - 4 | 389 | 67 | ||
FQ224857.1 | - | 3 - 4 | 417 | 67 | ||
X71873.1 | - | 3 - 4 | 381 | 67 | ||
X75253.1 | - | 3 - 4 | 406 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
