Assay Details
Gene Symbol: Slc25a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017307.3 | NP_059003.1 | 5 - 6 | 639 | 100 | |
GenBank mRNA | BC100077.1 | - | 5 - 6 | 623 | 7126394 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
