Assay Details
Gene Symbol: Mapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017347.2 | NP_059043.1 | 4 - 5 | 692 | 63 | |
GenBank mRNA | AF155236.1 | - | 4 - 5 | 668 | 63 | |
M38194.1 | - | 4 - 5 | 629 | 63 | ||
M61177.1 | - | 4 - 5 | 650 | 63 | ||
S46779.1 | - | 4 - 5 | 692 | 63 | ||
U12008.1 | - | 4 - 5 | 629 | 63 | ||
X65198.1 | - | 4 - 5 | 692 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
