Assay Details
Gene Symbol: Arpc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019289.2 | NP_062162.1 | 2 - 3 | 151 | 62 | |
GenBank mRNA | AF083269.1 | - | 1 - 2 | 64 | 62 | |
BC062027.1 | - | 2 - 3 | 151 | 5622004 | 62 | |
FQ228791.1 | - | 2 - 3 | 122 | 62 | ||
FQ228917.1 | - | 2 - 3 | 167 | 62 | ||
FQ228971.1 | - | 2 - 3 | 136 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
