Assay Details
Gene Symbol: Eif5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020075.1 | NP_064460.1 | 4 - 5 | 663 | 151 | |
XM_006240608.1 | XP_006240670.1 | 6 - 7 | 926 | 151 | ||
XM_006240609.2 | XP_006240671.1 | 5 - 6 | 1206 | 151 | ||
GenBank mRNA | BC062398.1 | - | 5 - 6 | 851 | 151 | |
L11651.1 | - | 4 - 5 | 663 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
