Assay Details
Gene Symbol: Ensa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033974.2 | NP_001029146.1 | 1 - 2 | 195 | 85 | |
NM_021842.3 | NP_068614.1 | 1 - 2 | 195 | 85 | ||
GenBank mRNA | AJ005984.1 | - | 1 - 2 | 55 | 85 | |
BC059135.1 | - | 1 - 2 | 199 | 85 | ||
CB812628.1 | - | 1 - 2 | 174 | 85 | ||
FQ212889.1 | - | 1 - 2 | 160 | 85 | ||
FQ212980.1 | - | 1 - 2 | 177 | 85 | ||
FQ213096.1 | - | 1 - 2 | 162 | 85 | ||
FQ213912.1 | - | 1 - 2 | 179 | 85 | ||
FQ214207.1 | - | 1 - 2 | 173 | 85 | ||
FQ216114.1 | - | 1 - 2 | 179 | 85 | ||
FQ216323.1 | - | 1 - 2 | 154 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
