Assay Details
Gene Symbol: Hspd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022229.2 | NP_071565.2 | 9 - 10 | 1279 | 128 | |
XM_006244932.2 | XP_006244994.1 | 9 - 10 | 1315 | 128 | ||
GenBank mRNA | BC086507.1 | - | 9 - 10 | 1279 | 128 | |
X53585.1 | - | 8 - 9 | 1143 | 128 | ||
X54793.1 | - | 8 - 9 | 1227 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
