Assay Details
Gene Symbol: Crip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022501.1 | NP_071946.1 | 2 - 3 | 258 | 72 | |
XM_008764943.1 | XP_008763165.1 | 2 - 3 | 522 | 72 | ||
GenBank mRNA | BC061774.1 | - | 2 - 3 | 227 | 5621903 | 72 |
D17512.1 | - | 2 - 3 | 258 | 72 | ||
FQ226314.1 | - | 2 - 3 | 266 | 72 | ||
FQ227529.1 | - | 2 - 3 | 268 | 72 | ||
FQ230331.1 | - | 2 - 3 | 268 | 72 | ||
FQ233584.1 | - | 2 - 3 | 272 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764943.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
