Assay Details
Gene Symbol: Pfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022511.2 | NP_071956.2 | 1 - 2 | 253 | 58 | |
GenBank mRNA | BC062405.1 | - | 1 - 2 | 253 | 58 | |
FQ217736.1 | - | 1 - 2 | 287 | 58 | ||
FQ221987.1 | - | 1 - 2 | 287 | 58 | ||
FQ227505.1 | - | 1 - 2 | 288 | 58 | ||
FQ228171.1 | - | 1 - 2 | 287 | 58 | ||
FQ232047.1 | - | 1 - 2 | 286 | 58 | ||
FQ232161.1 | - | 1 - 2 | 243 | 58 | ||
FQ232713.1 | - | 1 - 2 | 257 | 58 | ||
FQ232749.1 | - | 1 - 2 | 252 | 58 | ||
FQ235292.1 | - | 1 - 2 | 288 | 58 | ||
X96967.1 | - | 1 - 2 | 136 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
