Assay Details
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271057.1 | NP_001257986.1 | 13 - 14 | 1602 | 65 | |
NM_022516.5 | NP_071961.2 | 12 - 13 | 1524 | 65 | ||
GenBank mRNA | BC061858.1 | - | 13 - 14 | 1597 | 65 | |
X60789.1 | - | 12 - 13 | 1487 | 65 | ||
X60790.1 | - | 9 - 10 | 973 | 65 | ||
X74565.1 | - | 13 - 14 | 1583 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
