Assay Details
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271057.1 | NP_001257986.1 | 9 - 10 | 1187 | 73 | |
NM_022516.5 | NP_071961.2 | 8 - 9 | 1109 | 73 | ||
GenBank mRNA | BC061858.1 | - | 9 - 10 | 1182 | 73 | |
CK365398.1 | - | 5 - 6 | 695 | 73 | ||
X60789.1 | - | 8 - 9 | 1072 | 73 | ||
X60790.1 | - | 5 - 6 | 558 | 73 | ||
X74565.1 | - | 9 - 10 | 1168 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
