Assay Details
Gene Symbol: Gabarapl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022706.2 | NP_073197.1 | 3 - 4 | 326 | 120 | |
GenBank mRNA | AB003515.1 | - | 3 - 4 | 364 | 120 | |
BC058145.1 | - | 3 - 4 | 329 | 6921109 | 120 | |
BC088139.1 | - | 3 - 4 | 326 | 7323806 | 120 | |
FQ216852.1 | - | 3 - 4 | 340 | 120 | ||
FQ226397.1 | - | 3 - 4 | 350 | 120 | ||
FQ231643.1 | - | 3 - 4 | 319 | 120 | ||
FQ233551.1 | - | 3 - 4 | 361 | 120 | ||
FQ234861.1 | - | 3 - 4 | 319 | 120 |
Gene Symbol: LOC100359937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729947.4 | XP_002729993.3 | 1 | 315 | 120 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
