Assay Details
Gene Symbol: Prmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024363.1 | NP_077339.1 | 4 - 5 | 358 | 91 | |
XM_006229144.2 | XP_006229206.1 | 5 - 6 | 476 | 91 | ||
XM_006229145.2 | XP_006229207.1 | 4 - 5 | 640 | 91 | ||
XM_006229146.2 | XP_006229208.1 | 5 - 6 | 743 | 91 | ||
GenBank mRNA | BC078815.1 | - | 4 - 5 | 363 | 91 | |
FQ213198.1 | - | 4 - 5 | 371 | 91 | ||
FQ219843.1 | - | 4 - 5 | 369 | 91 | ||
FQ220982.1 | - | 2 - 3 | 110 | 91 | ||
FQ228625.1 | - | 4 - 5 | 639 | 91 | ||
FQ233897.1 | - | 4 - 5 | 371 | 91 | ||
U60882.1 | - | 4 - 5 | 358 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
