Assay Details
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030849.1 | NP_110476.1 | 1 - 2 | 73 | 78 | |
GenBank mRNA | S75359.1 | - | 1 - 2 | 73 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_030849.1, S75359.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
