Assay Details
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030849.1 | NP_110476.1 | 3 - 4 | 454 | 89 | |
GenBank mRNA | D17667.1 | - | 2 - 3 | 229 | 89 | |
D38082.1 | - | 2 - 3 | 311 | 89 | ||
S75359.1 | - | 3 - 4 | 454 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
